Sampling swabs
PP/PE
Katalog # PMPC25-806-1PD
Leverantör: PURITAN MEDICAL
Gå till:
Product Details & Documents
This synthetic tipped applicator is made of medical-grade polyester fibre that is carded in our factory to maintain consistency.
- Standard tip with medical grade quality
- Non-absorbent no fatty acids or latent sugars for specimen transport, lab work, and diagnostic kits
- Bonded securely to a semi-flexible PS handle by an aqueous based adhesive
Applications include diagnostic specimen collection, patient care, cell collection for DNA testing and gynaecological screening.
: 10 boxes per case of 100 individually wrapped packages.
: *Produkten/produkterna på denna sida ligger utanför tillämpningsområdet för EU -direktivet 2019/904
Specifikationer
- Shaft/head:PP/PE
- UN No.:3077
- Cat. No.:EHERC16630000
- CAS Nr.:119-12-0
- GHS-Signal:Achtung
- Molekulargewicht:340,33
- Beschreibung Gefahrenklasse:GHS07, GHS09
- Lagerbedingungen:4 °C
- Synonym(s):Phosphorothioic acid, O,O-diethyl ester, O-ester with 6-hydroxy-2-phenyl-3(2H)-pyridazinone (7CI,8CI);3(2H)-Pyridazinone, 6-hydroxy-2-phenyl-, O-ester with O,O-diethyl phosphorothioate (8CI);American Cyanamid 12,503;CL 12503;ENT 23,968;O,O-Diethyl O-(2,3-dihydro-3-oxo-2-phenyl-6-pyridazinyl) phosphorothioate;O,O-Diethyl phosphorothioate O-ester with 6-hydroxy-2-phenyl-3(2H)-pyridazinone;O-(1,6)-Dihydro-6-oxo-1-phenylpyridazin-3-yl) O,O-diethyl phosphorothioate;Ofunack;Oreste;Pyridafenthion;Pyridaphenthion;Phosphorothioic acid, O-(1,6-dihydro-6-oxo-1-phenyl-3-pyridazinyl) O,O-diethyl ester
- Shipping conditions:RT
- Formel:C₁₄H₁₇N₂O₄PS