Test kits, photometric, Water hardness, residual, LCK cuvette tests
Katalog # 705-3041
Leverantör: Hach
CAS-nummer:
Specifikationer
- Kemisk metod:Metalphthalein
- Förp.:24 Tests
- Beskrivning:Hardness (residual Ca + Mg)
- Seq:235
- Typ:LCK427
- För apparat:DR3900, DR6000, DR1900, DR2800, DR3800, DR5000
- Mätområde:0,02 - 0,6 °dH
0,1 - 2 mg/l Ca
Specifikationer
Om denna produkt
Pre-dosed reagents for maximum safety.
- Precise and reliable measurement values
- Maximum safety for users, thanks to the closed system and low amounts of reagents
- Convenient and error-free dosing of the reagents
- Barcode label for automatic recognition in the photometer
- Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C