HPLC columns, Hibar®, PharmPrep® P, RP-18e
Leverantör: MERCK PRODUCTION CHEMICALS
Certifikat
Om denna produkt
These ready to use columns are suitable for preparative chromatography.
- Spherical high-performance sorbent
Specifikationer
- Catalog No:
- 1.20573.0001
- 1.20574.0001
- Max. tryck:
- ≤ 25 bar
- ≤ 10 bar