Surgical blade, No. 3, stitch cutter
Katalog # SWAN0326
Leverantör: SWANN MORTON
Specifikationer
- Typ:No. 3
- Beskrivning:Stygnkniv, rostfritt stål, steril
- Förp.:100
Specifikationer
Om denna produkt
The stainless steel No. 3 or 'baby' stitch cutter is used in conjunction with the No. 3 surgical handle.
- Convenient and cost effective
- Sterile
Enables a more aseptic techinique by operating further away from the actual wound which also allows better visibility.
Fits handles 3 Graduated, 3L, 5B, 7, 9, B3 and B3L.