VWR®, Double-Frosted Microscope Slides
Catalog # 631-1161
Supplier: VWR Collection
Specifications
- Beskrivning:Double-frosted slides, ground edges, clipped corner 45°
- L×B:76×25 mm
- Tjocklek:1,0 (-0,05/+0,1) mm
- Förp.:2.500
Specifications
About this item
Microscope slides with double-frosted ends designed for human blood, body fluids tests, as well as paraffin sections.
- Shelf life of 60 months
- Smooth edges for operational safety
- Possible marking with pencils, marker pens and labelling systems
Suitable for use in haematology, microbiology, as well as histopathology applications.
Packaging: 50 pieces per box. 50 boxes per case.
Caution: Fragile items should be stored in dry and proper laboratory temperature conditions.