ContactUsLinkComponent
VWR® Closed Top PP Screw Caps for Storage Vials
VWR® Closed Top PP Screw Caps for Storage Vials
Katalog # 548-0901
Leverantör:  VWR Collection
undefined
VWR® Closed Top PP Screw Caps for Storage Vials
Katalog # 548-0901
Leverantör:  VWR Collection
Specifikationer
  • Hårdhet:
    55° shore A
  • Septum:
    Butyl röd/PTFE grå
  • Typ:
    PP vit, utan hål, för 548-3231
  • Seq:
    65
  • Tjocklek:
    1,3 mm
  • Förp.:
    1.000