Cleanroom notebooks, BCR®
Supplier: BERKSHIRE
Certificates
About this item
BCR MyNotes Notebooks are made with Berkshire`s BCR Bond cleanroom paper to minimise particle contamination. They are practical for note taking and data recording in the cleanroom. Bond is made with the purest quality cellulose from specially selected sustainable forests and is coated with a proprietary formula to minimise particle and fibre release. The cleanroom paper is recommended for use in ISO Class 3-7 cleanrooms.
- Compatible with gamma irradiation for use in sterile applications
- Compatible with standard copiers and printers
- Full lot tracking on every case
- Double packed for cleanliness
- Meets RoHS guideline
A4 or A5 size