Mesh woven filters, Propyltex
Catalog # SEFA05-150/34
Supplier: SEFAR
Specifications
- Porstorlek:150,00 µm
- Seq:02
- Förp.:1
Specifications
About this item
Precision woven synthetic monofilament fabrics. It’s a highly specialised monofilament fabric, characterised by precisely defined and controlled, consistent and repeatable material properties such as pore size, thickness, tensile strength, dimensional stability, cleanliness etc.
- Surface properties are even and consistent
- Thickness tolerance is within microns through entire rolls and lots
- Cleanliness meets strict standards