TLC imaging system, MultiDoc-It™
733-2048
- Beskrivning:TLC imaging system, MultiDoc-It™ 130
- Seq:01
- Förp.:1
For imaging in thin layer chromatography (TLC) applications, with mega pixel resolution digital colour camera for capturing gels, plates and membranes.
- Sturdy metal construction with large access door
- Hood enclosure provides a darkroom environment and protects the Digi camera
- Exclusive capture software enables image acquisition, enhancement and report capabilities
- Built-in white light; safety switch turns UV off when the door is open
- W×D×H: 356×323×483 mm
: MultiDoc-It™ TLC model includes two UVGL-25 UV lamps for placement on two sides of the darkroom. The system is run from a suitable laptop or PC which is not supplied. Please contact us for details on accessories, including printers, converter places, image analysis software.