End point detection system, End-Zone™
Leverantör: LABCONCO
Certifikat
Om denna produkt
End-Zone™ provides an alternative to visual inspection making it easy to determine when freeze drying is complete.
- Alerts when sample reaches the user-selected moisture level
- Works with FreeZone Lyo-Works™ OS
Starter kit is used for detecting end point in one sample flask. Add-on Kit is used for detecting end point in up to five sample.
End-Zone™ will alert when sample moisture levels have reached the user-selected level. It compares the vacuum level of an empty valve to the vacuum level inside a fast-freeze flask containing the sample.
Delivery: Starter Kit includes two vacuum sensors, one valve adapter, one fast-freeze flask top with sensor port, a connection cable, and a mountable holder with space for six sensors. Add-on kit includes one vacuum sensor and one Fast-Freeze flask top with an opening to insert the vacuum sensor.
Specifikationer
- Catalog No:
- 535-0392
- LBCP780902000
- Environmentally Preferable:
- Sustainable Packaging