Dehydrated culture media, china blue lactose agar
Katalog # SIAL22520-500G
Leverantör: Merck
CAS-nummer:
Specifikationer
- Förp.:500 g
- Standard:-
- Beskrivning:China blue lactose agar
Specifikationer
Om denna produkt
Selective medium for differentiating lactose fermenting and lactose non-fermenting microorganisms.