Sterilising box for petri dishes
Catalog # 216-0058
Supplier: BOCHEM
Specifications
- Höjd:250 mm
- Förp.:1
Specifications
About this item
Sterilising box made of stainless steel, for up to ten Petri dishes with Ø 100 mm.
Sterilising box made of stainless steel, for up to ten Petri dishes with Ø 100 mm.