VWR® pHenomenal®, Redoxelektrod
662-1165
- Anslutning:BNC, 1 m kabel
- Beskrivning:Redoxelektrod pHenomenal® ORP220, påfyllbar
- Material:Glas
- Elektrolyt:3 M KCl
- Förp.:1
Redox electrode with BNC connector and cable.
: Includes pH-electrode only.