Orders LinkContactUsLinkComponent
Indicator strips, non bleeding
Indicator strips, non bleeding
Indicator strips, non bleeding, 0 - 6,0
Katalog # 315052J
Leverantör:   VWR Chemicals
Indicator strips, non bleeding
Katalog # 315052J
Leverantör:  VWR Chemicals
Indicator strips, non bleeding
Indicator strips, non bleeding
Indicator strips, non bleeding, 0 - 6,0
Price
210,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 100 Items
Product Details & Documents
Non bleeding special indicator dyes that are covalently bound onto the reagent paper and then adhered to plastic strips.
Specifikationer
  • pH-område:
    0 - 6,0
  • Beskrivning:
    Indicator strips, non bleeding