Vattenprovtagare, SP 5 S-serien
Supplier: MAXX
Certificates
About this item
The SP 5 are modular fixed site units available in different configurations.
- Thermostatic control of the sample compartment to 4 °C with overheating protection and auto defrost
- Microprocessor to control and select sampling and bottle filling intervals with real time events
- Diaphragm pump with metering vessel made of glass Duran 50
Provtagarna ger justerbara, exakta provvolymer för uppsamling i en eller flera behållare.
Specifications
- Catalog No:
- 701-0144
- 701-0151
- 701-0104
- 701-0102
- 701-0139
- 701-0154
- 701-0150
- 701-0153
- 701-0098
- 701-0147
- 701-0101
- 701-0148
- 701-0149
- 701-0152
- 701-0141
- 701-0075
- 701-0146
- 701-0142
- 701-0100
- 701-0143
- 701-0145
- 701-0155
- 701-0105
- 701-0140
- 701-0103
- 701-0099
- Modell:
- SP5 S
- SP5 B
- SP5 S
- SP5 B
- SP5 B
- SP5 B
- SP5 S
- SP5 S
- SP5 S
- SP5 B
- SP5 S
- SP5 S
- SP5 S
- SP5 S
- SP5 S
- SP5 B
- SP5 S
- Provstorlek:
- 20 – 350 ml
- 20 – 350 ml
- 20 – 350 ml
- 20 – 350 ml
- 20 – 350 ml
- 20 – 350 ml
- 20 – 350 ml
- 20 – 350 ml
- 20 – 350 ml
- 20 – 350 ml
- 20 – 350 ml
- 20 – 350 ml
- 20 – 350 ml
- 20 – 350 ml
- 20 – 350 ml
- 20 – 350 ml
- Program:
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- 12
- Dataloggning:
- 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
- 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
- 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
- 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
- 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
- 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
- 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
- 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
- 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
- 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
- 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
- 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
- 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
- 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
- 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
- 3000 entries – Non volatile data memory ; Storage of sampling and malfunction data like sample extractions, bottle changes, messages, external signals – Optional with I/O add-on-board expandable to 32 GB.
- Noggrannhet:
- 2,8 %
- 2,8 %
- 2,8 %
- 2,8 %
- 2,8 %
- 2,8 %
- 2,8 %
- 2,8 %
- 2,8 %
- 2,8 %
- 2,8 %
- 2,8 %
- 2,8 %
- 2,8 %
- 2,8 %
- 2,8 %
- 2,8 %
- Signalingång:
- 2x analogue; 8x digital
- 2x analogue; 8x digital
- 2x analogue; 8x digital
- 2x analogue; 8x digital
- 2x analogue; 8x digital
- Signalutgång:
- Up to 8x digital
- Up to 8x digital
- Up to 8x digital
- Up to 8x digital
- Up to 8x digital
- Kommunikation:
- Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
- Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
- GSM-modem som tillval
- Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
- Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
- Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
- Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
- Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
- Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
- Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
- Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
- Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
- Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
- Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
- Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
- Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
- Optional Modbus connection via DP PROFIBUS ; LAN/WLAN via TCP/IP RJ45, with IE-Browser, memory 4-32GB SD/SDHC
- Utgång:
- Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
- Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
- Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
- Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
- Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
- Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
- Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
- Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
- Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
- Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
- Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
- Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
- Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
- Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
- Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
- Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
- Mini-USB, RS 422/485, RS 232 and optional Ethernet RJ45
- Kontroll:
- Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
- Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
- Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
- Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
- Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
- Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
- Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
- Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
- Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
- Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
- Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
- Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
- Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
- Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
- Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
- Microprocessed sleeping mode from 5 mA, foil keyboard and backlit graphical display (128*64 P)
- Sughöjd:
- <8 mWC
- <8 mWC
- <8 mWC
- <8 mWC
- <8 mWC
- <8 mWC
- <8 mWC
- <8 mWC
- <8 mWC
- <8 mWC
- <8 mWC
- <8 mWC
- <8 mWC
- <8 mWC
- <8 mWC
- <8 mWC
- <8 mWC
- Nätspänning:
- 8 – 16 V
- 8 – 16 V
- 8 – 16 V
- 8 – 16 V
- 8 – 16 V
- 8 – 16 V
- 8 – 16 V
- 8 – 16 V
- 8 – 16 V
- 8 – 16 V
- 8 – 16 V
- 8 – 16 V
- W×D×H:
- 690×645×1290 mm
- 760×745×1100 mm
- 690×645×1290 mm
- 760×745×1100 mm
- 760×745×1100 mm
- 760×745×1100 mm
- 690×645×1290 mm
- 690×645×1290 mm
- 690×645×1290 mm
- 760×745×1100 mm
- 690×645×1290 mm
- 690×645×1290 mm
- 690×645×1290 mm
- 690×645×1290 mm
- 690×645×1290 mm
- 760×745×1100 mm
- 690×645×1290 mm
- Vikt:
- 100 kg
- 75 kg
- 100 kg
- 75 kg
- 75 kg
- 75 kg
- 100 kg
- 100 kg
- 100 kg
- 75 kg
- 100 kg
- 100 kg
- 100 kg
- 100 kg
- 100 kg
- 75 kg
- 100 kg