Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Buffertar
pH buffertar
Buffers, concentrated solutions, Fluka™
Buffers, concentrated solutions, Fluka™
:
Honeywell Chemicals
Buffers, concentrated solutions, Fluka™
:
Honeywell Chemicals
sds_38740
sds_38746
sds_38752