Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Immunoglobulin Superfamily Containing Leucine-rich Repeat 2
- Antigensymbol:ISLR2
- Klonalitet:Polyclonal
- Konjugation:FITC (Fluorescein Isothiocyanate)
- Värd:Rabbit
- ImmunKemi:Yes
- Isotyp:IgG
- Reaktivitet:Human,Rat,Mouse
- Korsad adsorption:No
- Form:liquid
- Gen-ID:57611
- Antigen synonymer:ISLR2_HUMAN.|Immunoglobulin superfamily containing leucine-rich repeat protein 2|Leucine-rich repeat domain and immunoglobulin domain-containing axon extension protein
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Förvaringstemperatur:Store at –20 °C for 12 months
- Koncentration:1 μg/μl
- Shipping temperature:4°C
- Rening:Purified by Protein A
- Förp.:100 µl
Specifikationer
Om denna produkt
The leucine-rich (LRR) repeat is a 20-30 amino acid motif that forms a hydrophobic å/∫ horseshoe fold, allowing it to accommodate several leucine residues within a tightly packed core. All LRR repeats contain a variable segment and a highly conserved segment, the latter of which accounts for 11 or 12 residues of the entire LRR motif. ISLR2 (immunoglobulin superfamily containing leucine-rich repeat 2), also known as LINX (leucine-rich repeat domain and immunoglobulin domain-containing axon extension protein), is a 745 amino acid single-pass membrane protein that contains five LRR repeats, one Ig-like (immunoglobulin-like) domain, a LRRCT domain and one LRRNT domain. ISLR2 exists as a homomultimer and is essential for axon extension during neural development. The gene encoding ISLR2 maps to human chromosome 15q24.1.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ISLR2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat