Order Entry
Sweden
ContactUsLinkComponent
Anti-ISLR2 Rabbit Polyclonal Antibody (Cy7®)
Anti-ISLR2 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-9025R-CY7
Supplier:  Bioss
Anti-ISLR2 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-9025R-CY7
Supplier:  Bioss

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Immunoglobulin Superfamily Containing Leucine-rich Repeat 2
  • Antigensymbol:
    ISLR2
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Cy7®
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    57611
  • Antigen synonymer:
    ISLR2_HUMAN.|Immunoglobulin superfamily containing leucine-rich repeat protein 2|Leucine-rich repeat domain and immunoglobulin domain-containing axon extension protein
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at –20 °C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

Specifications

About this item

The leucine-rich (LRR) repeat is a 20-30 amino acid motif that forms a hydrophobic å/∫ horseshoe fold, allowing it to accommodate several leucine residues within a tightly packed core. All LRR repeats contain a variable segment and a highly conserved segment, the latter of which accounts for 11 or 12 residues of the entire LRR motif. ISLR2 (immunoglobulin superfamily containing leucine-rich repeat 2), also known as LINX (leucine-rich repeat domain and immunoglobulin domain-containing axon extension protein), is a 745 amino acid single-pass membrane protein that contains five LRR repeats, one Ig-like (immunoglobulin-like) domain, a LRRCT domain and one LRRNT domain. ISLR2 exists as a homomultimer and is essential for axon extension during neural development. The gene encoding ISLR2 maps to human chromosome 15q24.1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ISLR2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat