Order Entry
Sweden
ContactUsLinkComponent
Anti-EFHD1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anti-EFHD1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
  BOSSBS-9015R-A488
 :  Bioss
Anti-EFHD1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
  BOSSBS-9015R-A488
 :  Bioss

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    EF-hand Domain Family, member D1
  • Antigensymbol:
    EFHD1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 488
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    80303
  • Antigen synonymer:
    MST133|MSTP133|EF-hand domain-containing protein D1|SWS2|PP3051|EF-hand domain-containing protein 1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at –20 °C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

 

 

The EF-hand domain is a twelve amino acid loop motif that is commonly found in proteins that participate in calcium-binding events within the cell. EF-hand domains generally exist in a pair that, together, form a stable four-helix bundle that enables the binding of calcium ions. Swiprosin-2, also known as EFHD1 (EF-hand domain-containing protein D1), SWS2, PP3051 or MST133, is a 239 amino acid protein that contains two EF-hand domains and is expressed in a wide variety of tissues, including brain, liver, heart, kidney, testis, ovaries and spleen. Expression of Swiprosin-2, a possible calcium-binding protein, is upregulated during neuronal differentiation, suggesting a role for Swiprosin-2 in brain development and maturation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EFHD1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat