Order Entry
Sweden
ContactUsLinkComponent
Anti-GTDC1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-GTDC1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Katalog # BOSSBS-8582R-HRP
Leverantör:  Bioss
Anti-GTDC1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Katalog # BOSSBS-8582R-HRP
Leverantör:  Bioss

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Glycosyltransferase-like Domain Containing 1
  • Antigensymbol:
    GTDC1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    HRP (Horseradish Peroxidase)
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    79712
  • Antigen synonymer:
    GTDC1_HUMAN.|Mannosyltransferase candidate|Glycosyltransferase like 1|Glycosyltransferase like domain containing 1|Glycosyltransferase like domain containing protein 1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at –20 °C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

Specifikationer

Om denna produkt

Glycosyltransferases that mediate the regio- and stereoselective transfer of sugars, such as the fucosyltransferases, determine cell surface-carbohydrate profiles, which is an essential interface for biological recognition processes. GTDC1 (Glycosyltransferase-like domain-containing protein 1), also known as Mat-Xa, is a 458 amino acid protein belonging to the glycosyltransferase 1 family. GTDC1 is ubiquitously expressed, with highest levels found in peripheral blood leukocytes, spleen, lung and testis.An extremely rare recessive genetic disorder, Alstré°‰ syndrome, is caused by mutations in the ALMS1 gene, which maps to chromosome 2.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: GTDC1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat