Order Entry
Sweden
ContactUsLinkComponent
 
Anti-TYMSOS Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-9675R-A647
 :  Bioss
Anti-TYMSOS Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-9675R-A647
 :  Bioss

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    TYMS opposite strand
  • Antigensymbol:
    TYMSOS
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 647
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    494514
  • Antigen synonymer:
    C18orf56
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at –20 °C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

 

 

Encoding over 300 genes, chromosome 18 contains about 76 million bases. Trisomy 18, or Edwards syndrome, is the second most common trisomy after Downs syndrome. Symptoms of Edwards syndrome include low birth weight, a variety of physical development defects, heart deformations and breathing difficulty. Translocation between chromosome 18 and 14 is the most common translocation in cancers, and occurs in follicular lymphomas. Niemann-Pick disease, hereditary hemorrhagic telangiectasia and erythropoietic protoporphyria are associated with chromosome 18. The TGFβ modulators, Smad2, Smad4 and Smad7 are encoded by chromosome 18. The C18orf56 gene product has been provisionally designated C18orf56 pending further characterization.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TYMSOS
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human