Order Entry
Sweden
ContactUsLinkComponent
Anti-Proteasome 19S S7 Rabbit Polyclonal Antibody (Cy5®)
Anti-Proteasome 19S S7 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-9349R-CY5
Supplier:  Bioss
Anti-Proteasome 19S S7 Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-9349R-CY5
Supplier:  Bioss

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Proteasome 19S S7
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Cy5®
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    5701
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at –20 °C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

Specifications

About this item

The 26S protease is involved in the ATP-dependent degradation of ubiquitinated proteins. The regulatory (or ATPase) complex confers ATP dependency and substrate specificity to the 26S complex. In case of HIV-1 infection, positive modulator of Tat-mediated transactivation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Proteasome 19S S7
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat