Order Entry
Sweden
ContactUsLinkComponent
Anti-CNOT8 Rabbit Polyclonal Antibody (Cy3®)
Anti-CNOT8 Rabbit Polyclonal Antibody (Cy3®)
Katalog # BOSSBS-9388R-CY3
Leverantör:  Bioss
Anti-CNOT8 Rabbit Polyclonal Antibody (Cy3®)
Katalog # BOSSBS-9388R-CY3
Leverantör:  Bioss

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    CCR4-NOT transcription complex, subunit 8
  • Antigensymbol:
    CNOT8
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Cy3®
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    9337
  • Antigen synonymer:
    hCAF1|CALIF|Caf1b|POP2|CAF1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at –20 °C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

Specifikationer

Om denna produkt

The CCR4-NOT complex is an evolutionarily conserved, multi-component complex known to be involved in transcription as well as mRNA degradation. Various subunits within the complex are involved in influencing nuclear hormone receptor activities. The CCR4-NOT complex is also involved in the regulation of Histone H3 lysine 4 methylation through a ubiquitin-dependent pathway that likely involves the proteasome. CNOT8 (CCR4-NOT transcription complex subunit 8), also known as CALIF or POP2, is a 292 amino acid protein that localizes to both the nucleus and the cytoplasm and functions as part of the CCR-NOT complex. Expressed ubiquitously, CNOT8 plays a role in transcriptional regulation for a diverse set of processes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CNOT8
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity: