Order Entry
Sweden
ContactUsLinkComponent
Anti-HAGHL Rabbit Polyclonal Antibody (Cy5.5®)
Anti-HAGHL Rabbit Polyclonal Antibody (Cy5.5®)
Katalog # BOSSBS-8414R-CY5.5
Leverantör:  Bioss
Anti-HAGHL Rabbit Polyclonal Antibody (Cy5.5®)
Katalog # BOSSBS-8414R-CY5.5
Leverantör:  Bioss

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    hydroxyacylglutathione hydrolase-like
  • Antigensymbol:
    HAGHL
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Cy5.5®
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    84264
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at –20 °C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

Specifikationer

Om denna produkt

The hydroxyacylglutathione hydrolase-like protein (HAGHL) is a 290 amino acid protein that belongs to the glyoxalase II family. HAGHL binds two zinc ions per subunit and acts as a hydrolase on ester bonds. The gene encoding HAGHL maps to human chromosome 16, which encodes over 900 genes in approximately 90 million base pairs, makes up nearly 3% of human cellular DNA and is associated with a variety of genetic disorders. The rare disorder Rubinstein-Taybi syndrome is associated with chromosome 16, as is Crohn's disease, a gastrointestinal inflammatory condition that may involve the NOD2 gene. An association with systemic lupus erythematosis and a number of other autoimmune disorders with the pericentromeric region of chromosome 16 has led to the identification of SLC5A11 as a potential autoimmune modifier.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HAGHL
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat