Order Entry
Sweden
ContactUsLinkComponent
Anti-AGGF1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anti-AGGF1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Katalog # BOSSBS-7509R-A555
Leverantör:  Bioss
Anti-AGGF1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Katalog # BOSSBS-7509R-A555
Leverantör:  Bioss

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Angiogenic Factor With G Patch And FHA Domains 1
  • Antigensymbol:
    AGGF1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 555
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    55109
  • Antigen synonymer:
    HSU84971|GPATCH7|HUS84971|VG5Q|GPATC7
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at –20 °C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    492-537/714
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

Specifikationer

Om denna produkt

Promotes angiogenesis and the proliferation of endothelial cells. Able to bind to endothelial cells and promote cell proliferation, suggesting that it may act in an autocrine fashion.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AGGF1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat