Order Entry
Sweden
ContactUsLinkComponent
Anti-ICAM1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-ICAM1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-4616R-A350
Supplier:  Bioss
Anti-ICAM1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-4616R-A350
Supplier:  Bioss

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Intercellular Adhesion molecule 1
  • Antigensymbol:
    ICAM1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 350
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    3383
  • Antigen synonymer:
    ICAM-1|major group rhinovirus receptor|human rhinovirus receptor|BB2|cell surface glycoprotein P3.58|intercellular adhesion molecule 1|P3.58
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at 4°C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    240-290/532
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

Specifications

About this item

ICAM proteins are ligands for the leukocyte adhesion protein LFA-1 (integrin alpha-L/beta-2). During leukocyte trans-endothelial migration, ICAM1 engagement promotes the assembly of endothelial apical cups through ARHGEF26/SGEF and RHOG activation. In case of rhinovirus infection acts as a cellular receptor for the virus.

Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: ICAM1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human