Order Entry
Sweden
ContactUsLinkComponent
Anti-FPGT Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anti-FPGT Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-13211R-A647
Supplier:  Bioss
Anti-FPGT Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-13211R-A647
Supplier:  Bioss

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Fucose-1-Phosphate Guanylyltransferase
  • Antigensymbol:
    FPGT
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 647
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    8790
  • Antigen synonymer:
    GFPP
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at 4°C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

Specifications

About this item

Guanylyltransferase enzymes transfer one molecule of GTP to another molecule and also function in the transfer of guanosine nucleotides to sugar molecules. The carbohydrate moieties that are generated are covalently attached to cell surfaces and are necessary to ensure a surface contour that satisfies a variety of physiological roles. L-fucose is an important sugar in complex carbohydrates that is frequently found on plant and mammalian N-linked glycans. FPGT (Fucose-1-phosphate guanylyltransferase), also known as GFPP (GDP-L-fucose pyrophosphorylase), is a 594 amino acid cytoplasmic protein that catalyzes the formation of GDP-L-fucose from L-fucose-1-phosphate and GTP. FPGT functions to reutilize the L-fucose that is produced uopn glycoprotein and glycolipid turnover.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FPGT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat