Order Entry
Sweden
ContactUsLinkComponent
Anti-ARL4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-ARL4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Katalog # BOSSBS-12523R-HRP
Leverantör:  Bioss
Anti-ARL4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Katalog # BOSSBS-12523R-HRP
Leverantör:  Bioss

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    ADP-Ribosylation Factor-Like 4
  • Antigensymbol:
    ARL4
  • Klonalitet:
    Polyclonal
  • Konjugation:
    HRP (Horseradish Peroxidase)
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    10124
  • Antigen synonymer:
    ADP-ribosylation factor-like protein 4A|ADP ribosylation factor like 4A|ARL4A_HUMAN.|ARL4A|ARL 4
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at 4°C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

Specifikationer

Om denna produkt

ADP-ribosylation factors (ARFs) are highly conserved guanine nucleotide-binding proteins that enhance the ADP-ribosyltransferase activity of cholera toxin. ARF抯 are important in eukaryotic vesicular trafficking pathways and activating phospholipase D. ARL4 (ADP-ribosylation factor-like protein 4A) is a member of the ARF-like protein (ARL) subfamily of small GTPases. It contains a C terminal nuclear localization signal (NLS) region that interacts with Importin-? ARL4 localizes to the nucleus and is found in a variety of tissues, but is predominantly expressed in spermatogonia and Sertoli cells. It is most closely related to ARL6 and ARL7. Unlike ARFs, ARL4 does not activate the cholera toxin ADP-ribosyltranferase. ARL4 may play a role in neurogenesis during embryonic development and somitogenesis in the early stages of adult spermatogenesis.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: ARL4A
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat