Order Entry
Sweden
ContactUsLinkComponent
 
Anti-ARL1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-12522R-A647
 :  Bioss
Anti-ARL1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
  BOSSBS-12522R-A647
 :  Bioss

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    ADP-Ribosylation Factor-Like 1
  • Antigensymbol:
    ARL1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 647
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    400
  • Antigen synonymer:
    ADP-ribosylation factor-like protein 1|ARFL1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at 4°C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

 

 

The ADP-ribosylation factor (ARF) protein family are structurally and functionally conserved members of the Ras superfamily of regulatory GTP-binding proteins (1–3). ARFs influence vesicle trafficking and signal transduction in eukaryotic cells (1–3). ARF-dependent regulatory mechanisms include the coordination of spectrin interactions with golgi membranes and the association of actin to the golgi via rho family-dependent G-protein localization (Rac, CDC42) and WASP/Arp2/3 complexes (3–7). Additionally, ARFs play a central role in maintenance of organelle integrity, assembly of coat proteins, and activation of phospholipase D (5–7). The ARF proteins are categorized as class I (ARF1, ARF2,and ARF3), class II (ARF4 and ARF5) and class III (ARF6); members of each class share a common gene organization (8,9). The human ARF6 gene maps to chromosome 7q22.1, contains five exons and four introns, and encodes a 175 amino acid protein (8,9).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARL1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat