Order Entry
Sweden
ContactUsLinkComponent
Anti-BRWD3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-BRWD3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-11811R-A350
Supplier:  Bioss
Anti-BRWD3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-11811R-A350
Supplier:  Bioss

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    BromoDomain And WD Repeat Domain Containing 3
  • Antigensymbol:
    BRWD3
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 350
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Antigen synonymer:
    Bromo domain containing protein disrupted in leukemia|BRODL|Bromodomain and WD repeat domain containing 3|Novel WD repeat domain protein|BRWD3_HUMAN.
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at 4°C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

Specifications

About this item

Members of the WD repeat protein family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis and gene regulation. BRWD3 (bromodomain and WD repeat-containing protein 3) is a 1,802 amino acid protein expressed in fetal liver and most adult tissues. Existing as five alternatively spliced isoforms, BRWD3 contains two bromo domains, nine WD repeats and is thought to play a role in transcription by modifying chromatin. Mutations in the gene encoding BRWD3 are the cause of mental retardation X-linked type 93 (MRX93), which is also known as mental retardation X-linked with macrocephaly (XLMR). MRX93 is characterized by mild intellectual disability, macrocephaly, a prominent forehead and large cupped ears.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BRWD3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat