VWR® Beakers, Low Form, PMP (TPX)
Katalog #
Leverantör: VWR Collection
Leverantör #:
Specifikationer
- Catalog No:
- 213-0485
- 213-0484
- 213-0487
- 213-0486
- 213-0482
- 213-0510
- 213-0483
- 213-0488
- Environmentally Preferable:
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
- Sustainable Packaging
Specifikationer
Om denna produkt
Made of PMP (TPX), crystal-clear.
- Excellent chemical resistance
- Printed graduations
- Withstand temperatures of 0 to 121 °C when used continuously (up to 170 °C for short periods)
- Resin meets US FDA 21 CFR requirements