Medel mot sporer, Actril® medel för kallsterilisering
Katalog # 148-0115
Leverantör: MARCOR PURIFICATION
Specifikationer
- Beskrivning:Actril® CR, Cold Sterilant
- Förp.:1 l
Specifikationer
Om denna produkt
Actril® Cold Sterilant (Clean Room packaging) is a ready to use, effective, viricide, bactericide, fungicide, and sporicide It is is available in cleanroom packaging to make it easier to use in cleanroom setting. Actril® CR is completely and does not have to be part of a rotation of disinfectants program.
- No dilution required
- Faster, safer, and less corrosive than aldehyde-based liquids
- Filtered through 0,2 micron filters
- Double bagged and EtO Processed (6 log reduction of B. atrophaeus spores)
- Complete lot traceability and documentation conforms to the pharma standards
Actril® Cold Sterilant is a leading surface disinfectant in medical and cleanroom applications.
Certifications: Actril® Cold Sterilant complies with the USP guidelines for choosing disinfectants for aseptic areas.
Packaging: 6×1 kg bottles with sprayers per case.