ContactUsLinkComponent
VWR® Pipette Charging Stand, Rotary
VWR® Pipette Charging Stand, Rotary
Catalog # 613-5747
Supplier:  VWR Collection
undefined
VWR® Pipette Charging Stand, Rotary
Catalog # 613-5747
Supplier:  VWR Collection
Specifications
  • Färg:
    Vit
  • Beskrivning:
    Pipette charging stand, rotary, for up to 5 electronic multi and single channel pipettes
  • Förp.:
    1
Specifications
About this item
This power carousel is a comfortable charging solution for up to five electronic pipettes.
  • Protects valuable instruments from shocks and contamination
  • Pipettes hang vertically to prevent liquids from running inside the shaft
  • Rotating head gives users easy access to the pipette of their choice
  • Slim design allows users to have a neat, tidy and organised work bench
  • Unrollable cable will avoid any cable spaghetti issue