Capillary tubes with filament
Catalog # HARV30-0020
Supplier: HARVARD APPARATUS
Specifications
- Ø utv.:1,0 mm
- Ø inr.:0,58 mm
- Längd:150 mm
- Seq:05
- Förp.:500
Specifications
About this item
Capillary tubes, made of high quality borosilicate glass with filaments.
- Easy filling due to filament
- Available in standard or thin wall configurations
- Low tip impedance (less than 50 MΩ)
- Good for patch clamp pipettes and microinjection needles