Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Buffertar
pH buffertar
pH buffer solutions, referenced at 20 °C, traceable to SRM from NIST, Fluka™
pH buffer solutions, referenced at 20 °C, traceable to SRM from NIST, Fluka™
:
Honeywell Chemicals
pH buffer solutions, referenced at 20 °C, traceable to SRM from NIST, Fluka™
:
Honeywell Chemicals
Citrate and phosphate buffer solutions contain fungicide
Traceable to SRM from NIST and PTB
Documentation
SDS
sds_33646
sds_33648
Documentation