Vertical manifold for drying chambers
CHRS121226
- Beskrivning:Vertical manifold with 8 rubber valve connection, stainless steel, DIN external ground surface NS 45/40, for connecting vessels 8×100/250/500/1000 ml, 4×2000 ml round-bottom flask and 8×75/150/300/600/1200 ml, 4×2000 ml wide neck filter bottle, W×H: 380×755 mm
- Förp.:1
Stainless steel Manifold without cover: optional acrylic or glass cover.
- Condensate retention tank protects the freeze-dryer
- The connections can be shut off individually and the drying vessels can be vented individually