Weighing pans, aluminium
Catalog # KOEH2-1607
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Specifications
- Kapacitet:160 ml
- Färg:Silver
- Beskrivning:Rectangular aluminium shells with edge and conical tapering
- B×D×H:64×100×38 mm
- Förp.:100
Specifications
About this item
Made of Aluminium, bright.
- Good temperature resistance
- Good chemical resistance
- Lightweight
- Stackable