Ion exchange resins, Diaion® UBK550 (Na)
Catalog # 46609.A1
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Partikelstorlek:0,22 mm
- Förp.:1 kg
- Beskrivning:Ion exchange fractionation resin, 8% cross-linked, strongly acidic gel type, 1,9 meq/ml on PS-DVB
Specifications
About this item
These resins are ideal for use in chromatographic oligosaccharide sugar separations and sugar alcohol separations.