Industrial overalls, AlphaTec® 1500, model 138
: Ansell
These overalls are made from a triple layer SMS fabric that has been proven to filter 100% of particles larger than 3,0 µm (according to JSTIIF particle penetration test).
- Elasticated 3-piece hood, wrists, waist and ankles
- Two-way front zipper with re-sealable storm flap
- Stitched seams combine strength with particle barrier
- Air and water vapour permeable ('breathable') to help reduce the risk of heat stress
- Silicone-free: Critical in spray painting applications
- Optimised body fit improves wearer comfort and safety
Designed for workers involved in the stripping, clear up or handling of asbestos, general maintenance, construction and contract cleaning. Asbestos fibres, such as Chrysotile, are typically 3 to 5 µm in size.
Weight:
White: 48 g/m²
Dark blue/Red: 50 g/m²
: Type 5/6, EN 1073-2; Cat. III.
- Catalog No:
- ANSEWH15S-00138-06
- ANSEWH15S-00138-05
- ANSEWH15S-00138-04
- ANSEWH15S-00138-03
- ANSENV15S-00138-07
- ANSEWH15S-00138-02
- ANSENV15S-00138-06
- ANSENV15S-00138-05
- ANSENV15S-00138-04
- ANSENV15S-00138-03
- ANSEWH15S-00138-07
- Modell:
- 138
- 138
- 138
- 138
- 138
- 138
- 138
- 138
- 138
- 138
- 138
- Stängningsstorlek:
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Seam type:
- Taped
- Taped
- Taped
- Taped
- Taped
- Taped
- Taped
- Taped
- Taped
- Taped
- Taped
- Outfit applikationer:
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- General Purpose, Industrial
- Vikt:
- 48 / 50 g/m²
- 48 / 50 g/m²
- 48 / 50 g/m²
- 48 / 50 g/m²
- 48 / 50 g/m²
- 48 / 50 g/m²
- 48 / 50 g/m²
- 48 / 50 g/m²
- 48 / 50 g/m²
- 48 / 50 g/m²
- 48 / 50 g/m²