ContactUsLinkComponent
Scraper for foodstuff, SteriPlast®
Katalog # BURK5378-3030
Leverantör:  BURKLE
undefined
Scraper for foodstuff, SteriPlast®
Katalog # BURK5378-3030
Leverantör:  BURKLE
Specifikationer
  • Sterila:
    80 mm
  • Längd:
    200 mm
  • Förp.:
    10
Specifikationer
Om denna produkt
PS, blue. This scraper is suitable for scraping off, smoothing or cleaning as well as removing residue. Especially designed for use in food, animal feed and pharmaceutical industries.
  • Disposable
  • Sturdy and convenient
  • Produced in a class 7 cleanroom (10000)
  • Individually packaged and then sterilised by gamma rays
  • Can be used as part of HACCP/IFS foreign object management
The shade of blue that is used does not occur in a natural form in foodstuff. Because of this, this blue product or parts of it can be quickly found and easily identified in a visual check if they are ever mixed into production.