Sequencing septa mats, Axygen®
Katalog # 731-4130
Leverantör: Corning
Specifikationer
- Beskrivning:Matta för sekvensering, Septa Mat
- Förp.:10
Specifikationer
Om denna produkt
Silicone septa mat for capillary sequencing of 96-well PCR plates. Mat forms a tight seal on wells but is easily accessible through septa in each well. Ideal for work on ABI multi-capillary sequencing instruments.