Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Förvaring av tunnor
Accessories for drum filling station
Accessories for drum filling station
Accessories for drum filling station
Catalog # CEMG8675
Leverantör:
CEMO
Expand image
Accessories for drum filling station
Catalog # CEMG8675
Leverantör:
CEMO
Specifikationer
Vikt:
15 kg
Beskrivning:
Drum pallet, PE, for 2×200 l drums
B×D×H:
940×1380×390 mm
Förp.:
1
Specifikationer
Om denna produkt
Product Details
Product Details