Dosimeter för organiska ångor
Dosimeter för formaldehyd
514-0008
: 3M
Monitors use the proven scientific principle of diffusion, which involves the gradual mixing of molecules of two or more substances.
- Formaldehyde monitor uses a bisulphite-impregnated filter and is analysed by colorimetry
- Can be used for eight-hour TWA sampling
- Does not have the sensitivity for short-term exposure limit (STEL) monitoring
Designed for measuring personal exposure in work areas in a variety of industries.
18-month shelf life.
: Meet the OSHA requirement of ±25% accuracy at 0,5 - 5,0 ppm.
- Beskrivning:Dosimeter för formaldehyd
- Seq:02