S-protein agarose, Novagen
Katalog # 69704-4
Leverantör: Merck Millipore (Novagen)
CAS-nummer:
Specifikationer
- Partikelstorlek:45 - 165 µm
- Förp.:10 mL
- Beskrivning:S-Protein Agarose
Specifikationer
Om denna produkt
S-protein agarose specifically retains STag™ fusion proteins. The capacity of S-protein agarose varies and is based on the size and folding characteristics of a given target protein.
Binding can be performed in a column or batch mode. Elution is possible using slightly denaturing conditions or using a specific protease, such as thrombin or enterokinase, with appropriate recombinant proteins.