D-Desthiobiotin
Katalog # 71610-3
Leverantör: Merck Millipore (Novagen)
CAS-nummer:
Specifikationer
- Förpackningstyp:Plastic ampoule
- Förp.:1 g
- Beskrivning:D-Desthiobiotin
Specifikationer
Om denna produkt
D-Desthiobiotin provides a gentle elution of StrepTag® II proteins from the biotin-binding site of StrepTactin® resins. D-Desthiobiotin is offered as a lyophilised powder for preparation of elution buffer.