Glass microfibre filters with binder, grade GF 92, Whatman™
Supplier: Whatman products (Cytiva)
Certificates
About this item
Whatman Grade GF 92 glass microfiber filters from Cytiva are designed for prefiltration in order to protect against membrane fouling. Grade GF 92 is strengthened with an inorganic binder.
- Inorganic binder provides increased strength
- Temperature resistant up to 500 °C
- Medium filtration speed
Cytiva's business offers a variety of binder-strengthened glass microfiber filters for your filtration and prefiltration needs, including environmental, air pollution, water analysis.
Prefiltration can improve performance and longevity of equipment in applications such as:
- Measurement of crop protection agent using gas chromatography (GC) or HPLC
- Separation and determination of cold sludge when brewing beer
- Soot separation prior to use of gas analyzers
- Roll filters for automatic air filtration units
Specifications
- Catalog No:
- 514-3816
- 514-3817
- 514-3819
- 514-3818
- 514-3821
- 514-3814
- Klass:
- GF 92
- GF 92
- GF 92
- GF 92
- GF 92
- GF 92
- Vikt:
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- 70 g/m²
- Tjocklek:
- 350 µm
- 350 µm
- 350 µm
- 350 µm
- 350 µm
- 350 µm
- Luftflöde:
- 27 s/100 ml/1,56 cm²
- 27 s/100 ml/1,56 cm²
- 27 s/100 ml/1,56 cm²
- 27 s/100 ml/1,56 cm²
- 27 s/100 ml/1,56 cm²
- 27 s/100 ml/1,56 cm²
- Max. temperature:
- 500 °C
- 500 °C
- 500 °C
- 500 °C
- 500 °C
- 500 °C