Fluorometric drug discovery assay kit, Green SIRT5, Fluor-de-Lys®
Catalog # ENZOBMLAK5140001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Förp.:100 Tests
- Beskrivning:FLUOR DE LYS® Green SIRT5 fluorometric drug discovery kit
- Seq:02
Specifications
About this item
FLUOR DE LYS® Green SIRT5 fluorometric drug discovery kit is a sensitive fluorometric assay that measures the lysyl desuccinylase activity of the recombinant human SIRT5.
- Easy to use kit (two-step) for screening SIRT5 inhibitors/activators; includes enough active enzyme for entire plate
- Suitable for high-throughput screening (Z’-factors >0,73)
- Optimal, specific SIRT5 substrate means low enzyme concentration, making ‘hit’ validation easy
FLUOR DE LYS®-Succinyl Green substrate has longer excitation/emission wavelengths, avoiding interference often seen with screening compounds at shorter wavelengths.
Delivery: Kit contains SIRT5 (Sirtuin 5) (human, recombinant), FLUOR DE LYS®-Succinyl Green, Desuccinylase Substrate, FLUOR DE LYS® Developer Concentrate (20X), Nicotinamide (Sirtuin Inhibitor), Suramin sodium (Sirtuin inhibitor), FLUOR DE LYS®-Green Desuccinylated Standard, Sirtuin Assay Buffer, and half volume 96-well black NBS™ microplate.