Bordetella pertussis toxin
Katalog # ENZOBMLG1010050
Leverantör: ENZO LIFE SCIENCES
CAS-nummer:
Specifikationer
- Förp.:50 µG
- Beskrivning:Bordetella pertussis toxin, 50 µg
- Seq:02
Specifikationer
Om denna produkt
Bordetella pertussis toxin (Islet-activating protein, PTX) from Bordetella pertussis, strain NCTC 10739. is a high purity toxin applicable to ELISA, antibody capture, cytotoxicity studies, LINE assay.
- High purity, native protein available as >98% pure
- Minimal lot-to-lot variation ensures reproducible results
- Lyophilised format for extended shelf-life