Ion exchange resins, Diaion® HP20
Katalog # 46488.A1
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Partikelstorlek:0,25 mm
- Förp.:1 kg
- Beskrivning:Synthetic adsorbent ion exchange resin, Diaion® HP20, highly porous type
Specifications
About this item
Diaion® HP20 is used as a filler in chromatography columns due to its extremely large pores. It is considered as an effective adsorbent to eliminate toxic effects in fermentation and separation of traces heavy metal ions. It is also useful for the purification of peptides, proteins, fermentation products, polyphenols and cephalosporin C. It helps to remove bitter tasting substances from fruit juices.