Titrator, sample changer, AS1000
Katalog # 661-0247
Leverantör: Hach
Specifikationer
- Beskrivning:Sample changer, AS1000, 30 positions, 50 ml beaker, UK plug
- Seq:02
- Strömförsörjning:50/60 Hz
- Vikt:10 kg
- B×D×H:395×535×300 mm
- Förp.:1
Specifikationer
Om denna produkt
This automatic sample changer is designed to be used in analytical laboratories in connection with AT1000 Series titrators.
- Run sample analyses in total autonomy, without user intervention
- Rotating module turns the tray to change the sample position while the lifting module moves sensors and tubes up and down into each sample
- Automatic stirring of samples thanks to integrated magnetic stirrer
- Entirely PC-controlled with the TM1000 software to automate sample analysis series
The sample changer covers a wide variety of aqueous sample applications such as pH/conductivity/alkalinity and hardness in water, chlorides content in food products, pH/acidity in wine and juices, free and total SO₂ in wine.
Delivery: Supplied with lifting module, rotating module with carousel tray, support base with magnetic stirrer, probe and tubes holder, RS-232 cable with USB/Serial adapter to connect AS1000 to PC, power supply, set of magnetic stir bars, set of PP beakers and TM1000 PC-Software with ethernet cables to connect AT1000 to local network or PC.